Learn More
Kartell™ Pluripet II Variable Volume Micropipette
Click to view available options
Volume (Metric):
0.5, 5.0, 10.0 μL
100, 500, 1000 μL
Unit Size:
Each
Description
Specifications
Specifications
| Accuracy | ± 5,0 ± 2,5 ± 1,5 |
| Compatible Tips | Kartell™ 956 |
| Autoclavable | Autoclavable |
| Material | Plastic |
| Model | Pluripet II 10 |
| Product Line | Pluripet II |
| Type | Micropipette |
| Volume (Metric) | 0.5, 5.0, 10.0 μL |
| Increments | 0.1μL |