Learn More
Thermo Scientific™ Plunger-in-Needle Autosampler Syringes for Thermo Scientific™ Instruments
Product Code. p-4525907 Shop All Thermo Scientific Products
Change view
Click to view available options
Needle Gauge:
23 G
26 G
31 G
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Needle Gauge unitSize
10310975 31 G Each
12886036 23 G Each
10585245 26 G Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10310975 Supplier Thermo Scientific™ Supplier No. 36500500
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Get accurate and reproducible results over a range of volumes with Thermo Scientific™ Plunger-in-Needle Autosampler Syringes for Thermo Scientific Instruments.
Get accurate and reproducible results over a range of volumes with Thermo Scientific™ Plunger-in-Needle Autosampler Syringes for Thermo Scientific Instruments.
  • Manual syringes in industry-standard size
  • White scale backing for easy reading
  • Solvent-resistant markings
TRUSTED_SUSTAINABILITY
Specifications
Description 0.5uL volume; 31 gauge; 75mm length
Needle Gauge 31 G
Product Type GC Autosampler Syringe, Plunger-in-Needle
Disposable Reusable
Volume (Metric) 0.5 μL
Length (Metric) Needle 75 mm
Needle Point Style Cone
For Use With (Equipment) TriPlus, AS3000, AS2000, AS200, and AS800 Autosampler