Learn More
Memmert™ Plug-In Tube Extension for SF750, Straight
Product Code. 17598345 Shop All Memmert Products
Change view
Click to view available options
For Use With (Equipment):
SF750 Sterilizer
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
17598345 SF750 Sterilizer Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17598345 Supplier Memmert™ Supplier No. SF750B29719
Estimated Shipment: 05-11-2026
Plug-in tube extension designed for exhaust air ducting and connection by hose if necessary.
  • Accessory
  • Plug-in tube extension (outer diameter 60.3 mm, inner 57 mm)
  • Straight
Specifications
Diameter (Metric) Outside 60.3 mm
For Use With (Equipment) SF750 Sterilizer