Learn More
BRAND™ PLTconnect™ Software
Product Code. 15510682 Shop All Brand Products
Change view
Click to view available options
For Use With (Equipment):
PC USB port and Pipettes
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15510682 PC USB port and Pipettes Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15510682 Supplier BRAND™ Supplier No. 703980
  • ONLINE EXCLUSIVE PRICE
    163.00€
    152.21€ / Pack of 1
    Save 10.79€
    6% Off
Estimated Shipment: 09-12-2026
Software for PLT unit
  • The PLTconnect™ software gives you the additional security of knowing all your test results are documented at all times
  • PLTconnect™ transmits test results for each pipette automatically and securely through the USB port on the PC, storing them there in a database or as a test certificate
  • This means you always have access to test results for your pipettes, and can print out certificates as needed
TRUSTED_SUSTAINABILITY
Specifications
Description Automatically and securely transmit test results for each pipette
For Use With (Application) Documenting Test Results
For Use With (Equipment) PC USB port and Pipettes