Learn More
Platinum On Carbon (10 wt. %), TRC
Product Code. 30425940
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30425940 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30425940 Supplier Toronto Research Chemicals Supplier No. TRCP5780052.5G
 Restricted Item
Estimated Shipment: 14-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Platinum
CAS 7440-06-4
Molecular Formula Pt
Quantity 2.5 g
Synonym Pt
InChI Formula InChI=1S/Pt
SMILES [Pt]
IUPAC Name platinum
Molecular Weight (g/mol) 195.08
Formula Weight 194.965