Learn More
Thermo Scientific™ Accessories for Digital Microplate Shakers
Product Code. 16504802 Shop All Thermo Scientific Products
Click to view available options
Description:
96-well microplate clamps
Platform Large
Platform Large Assemble
Dimensions (L x W x H):
54 x 19 x 19 mm
292 x 275 x 14 mm
292 x 275 x 25 mm
Unit Size:
Each
Product Code. 16504802
Brand: Thermo Scientific™ 88882115
Includes
Pack of 7 Clamps
Get assorted accessories for Digital Microplate Shakers
Assorted accessories for Digital Microplate Shakers
Service Plan
Standard Thermo warranty guideline, provide repair service and free spare part within warranty
Warranty
2 years
TRUSTED_SUSTAINABILITY
Specifications
Product Type 96-well Microplate Clamps
Material Stainless Steel
Dimensions (L x W x H) 54 x 19 x 19 mm
Description 96-well microplate clamps
For Use With (Equipment) Rocker, Rotator, mixer
Includes 7 Clamps