Learn More
Eppendorf™ Platform for Innova™ 2000 Shaker and Erlenmeyer Flasks
Click to view available options
Holds:
10 mL Erlenmeyer Flasks
Unit Size:
Each
Description
- Holds 10 mL Erlenmeyer flasks
- Dedicated platform
- Fits Innova™ 2000 shaker model
- Made of stainless steel
Specifications
Specifications
| Holds | 10 mL Erlenmeyer Flasks |
| Material | Stainless Steel |
| Dimensions (L x W) | 33 x 28 cm |
| For Use With (Equipment) | Innova™ 2000 Shaker |
| No. of Positions | 60 |