Learn More
Thermo Scientific™ Plates and Modules with Affinity Binding Surfaces
Product Code. 10551493
Click to view available options
Color:
Black
Clear
White
Product Type:
Breakable Module F8 (x12) in Frame
Lock-Well Module C8 (x12) in Frame
Microplate
Solid Module F8 (x12) in Frame
Surface Treatment:
Glutathione
Streptavidin
Total Volume (Metric) Well:
120 μL
350 μL
400 μL
Unit Size:
Pack of 15
Pack of 5
Product Code. 10551493
Brand: Thermo Scientific™ 436018
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Help reduce variability in molecular orientation with these plates and modules with affinity binding surfaces.
Help reduce variability in molecular orientation with Thermo Scientific™ Plates and Modules with Affinity Binding Surfaces.
Affinity Capture Surfaces (Passive Streptavidin and Immobilizer Streptavidin, Nickel Chelate and Glutathione, BioBind)
  • Highly specific binding
  • Reduced variability in molecular orientation
  • Immobilizer surfaces improve signal to noise ratios
  • Streptavidin biotin interaction can be exploited to immobilize a wide range of biomolecules
TRUSTED_SUSTAINABILITY
Specifications
Detection Method Chemiluminescence
Packaging 15 Cases, 1 Pk.
Binding Type Affinity
Color White
Product Type Microplate
Material Polystyrene
Volume (Metric) Well 120 μL
No. of Wells 384
Volume (Metric) Working 50 μL
Surface Treatment Streptavidin
Total Volume (Metric) Well 120 μL
Well Shape Flat Bottom (F-well)
No. per Case 15
No. per Pack 1
Plate Blocking Unspecified
Target Molecule Biotinylated