Learn More
DWK Life Sciences Plate Holder, for glass ceramic laboratory protection plate
Ideal holder for glass ceramic plates for mounting on tripods.
Brand:  DWK Life Sciences 290785303
172.90 EUR valid until 2024-12-31
Use promo code "23258" to get your promotional price.
Additional Details : Weight : 1.07000kg
 View more versions of this product
Product Code. 11752909
  • 182.00€ / Pack of 5
Estimated Shipment: 23-07-2024
Description
In the laboratory, solutions are often heated. This step can be carried out using different techniques, e.g. in a water or sand bath, with a heating hood or classically using a Bunsen burner. If the Bunsen burner is used as the heat source, it is recommended to use a tripod or square foot and a corresponding glass ceramic plate. The vessel to be heated is placed on the glass ceramic plate with the Bunsen burner underneath. Contamination of the glass ceramic plate can be removed with the cleaning scraper.
  • Made from heat-resistant chrome-nickel steel with
Specifications
Specifications
Chrome-nickel Steel
135 mm
Plate holder for glass ceramic plates; Heating; Suitable for mounting on tripods
5/Pk.