Learn More
Kartell™ Plastilab™ Tapered Y-Shaped Tubing Connectors
BIOPHARMA
For use with flexible tubing.
Brand:  Kartell™ 7052700
Product Code. 12599746
  • ONLINE EXCLUSIVE PRICE
    12.00€
    11.02€ / Pack of 10
    Save 0.98€
    8% Off
 Restricted Item
In Stock
Edge
Description
  • Constructed from polypropylene
  • Autoclavable