Learn More
3M™ Plastic Sweatband
Product Code. 15729867
Change view
Click to view available options
For Use With (Equipment):
3M Headgear Combinations G500 Series
All 3M™ Hard Hats
Unit Size:
Pack of 10
Pack of 20
Pack of 250
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
15729867 3M Headgear Combinations G500 Series Pack of 10
15739957 All 3M™ Hard Hats Pack of 20
17267206 All 3M™ Hard Hats Pack of 250
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15729867 Supplier 3M™ Supplier No. 7000103789
  • ONLINE EXCLUSIVE PRICE
    53.00€
    41.85€ / Pack of 10
    Save 11.15€
    21% Off
Estimated Shipment: 20-10-2026
Add to basket
Add to basket
3M™ Plastic Sweatband suitable for use with 3M™ hard hats and headgear combinations.
  • Replacement plastic sweatband suitable for use with 3M™ Hard Hats and 3M™ G500 Headgear Combinations
  • Quick and easy to install
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance CE Approved
Color Black
For Use With (Equipment) 3M Headgear Combinations G500 Series
Material Plastic
Product Type Sweatband