Learn More
Fisherbrand™ Nylon Sieve Filter, 1m x 1m
Product Code. 11715488 Shop All Fisherbrand Products
Click to view available options
Pore Size:
5 μm
11 μm
20 μm
25 μm
30 μm
41μm
60 μm
70 μm
80 μm
100 μm
150 μm
200 μm
250 μm
300 μm
400 μm
500 μm
1000 μm
1250 μm
1400 μm
1550 μm
1700 μm
1800 μm
2000 μm
Unit Size:
Each
Product Code. 11715488
Only null left
Add to basket
Add to basket
For sieving or precision filtration of biological and chemical solutions. Fisherbrand™ Nylon Sieve Filters, 1m x 1m are available in a comprehensive range of pore sizes.
  • Resistant to aggressive solvents
  • Excellent compatibility with alcohols
  • Operating temperature of -40° to +115°C
  • Applications include solvent filtration and particle analysis
Specifications
Pore Size 20 μm
Frame Material Plastic
Mesh Material Nylon