Learn More
Sartorius Plastic Screw Box for Weights
Product Code. p-7228469 Shop All Sartorius Products
Click to view available options
Holds:
2 kg Weights
Unit Size:
Each
Product Code. 16333136
Brand: Sartorius YAW2000GL
Only null left
Add to basket
Add to basket
Plastic Screw Boxes for preventing any damage to the surface of weights.
  • Plastic screw boxes with foam inlet to be used for weights.
TRUSTED_SUSTAINABILITY
Specifications
Material Plastic
For Use With (Application) For use in cleanrooms
Holds 2 kg Weights