Learn More
Fisherbrand™ Plastic Hydrometer, 0-35% Brix
Click to view available options
Accuracy:
± 0.5 %mas/Brix
Unit Size:
Each
Description
- Made of shatter proof plastic for break resistance
- Made of HACCP compliant plastic that is suitable for use with food
- A saccharometer is a hydrometer for measuring the sugar content of a solution
- Scale division of 0.5 %mas/Brix, with accuracy of ± 0.5 %mas/Brix
- Instrument is single-packed in a transparent square plastic sleeve
Specifications
Specifications
| Type | Saccharometer/Brix Scale Hydrometer |
| Accuracy | ± 0.5 %mas/Brix |
| Increments | 0.5%mas/Brix |
| Certifications/Compliance | DIN EN ISO/IEC 17025 |
| Length (Metric) | 300 mm |
| Includes | Square plastic sleeve |
| Brix Range | 0 to 35° |
| Material | Plastic |
| For Use With (Application) | Measuring density of liquids/food, especially measuring sugar content |
| Temperature of Standardization | 20°C |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.