Learn More
Kappes Systeme™ Plastic Cross Divider
Product Code. 10644303
Encompass_Preferred
Change view
Click to view available options
Height (Metric):
85 mm
105 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (Metric) Unit Size
10644303 105 mm Each
10562553 85 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10644303 Supplier Kappes Systeme™ Supplier No. 6640.00.3457
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
Specifications
Product Type Cross Divider
Color Transparent
Material Plastic
For Use With (Equipment) Precision storage grance
Quantity 1/Pk.
Dimensions (L x W) 153 x 105 mm (WxH)
Height (Metric) 105 mm
Width (Metric) 153 mm