Learn More
Epredia™ Plastic Block Filing Cabinet
Product Code. 12513587 Shop All Epredia Products
Click to view available options
Product Type:
Plastic Block Filing Cabinet
Unit Size:
Each
Product Code. 12513587
Brand: Epredia™ 160000
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Simple filing of up to 1500 blocks. Epredia™ Plastic Block Filing Cabinets feature smooth sliding action and rigid construction.
  • Six drawers included
  • Total Capacity of 1500 blocks
  • Dimensions: 19.7 x 6.3 x 20.5 in. (50 x 16 x 52cm)
TRUSTED_SUSTAINABILITY
Specifications
Product Type Plastic Block Filing Cabinet
Holds 1500 Blocks
Depth (English) 19.7 in.
Depth (Metric) 50 cm
Height (English) 6.3 in.
Height (Metric) 16 cm
Width (English) 20.5 in.
Width (Metric) 52 cm