Learn More
BRAND™ Plastibrand™ PMP Flasks with Polypropylene Stopper
Shop All Brand Products
Click to view available options
Capacity (Metric):
10 mL
25 mL
Unit Size:
Each
Pack of 2
Description
- Tolerances according to class A / B, DIN EN ISO 1042.
- Calibrated to contain (TC, In).
- Exposure to temperatures up to 121°C (autoclaving) will not cause permanently exceeded error limits
- Cleaning up to max. 60°C is recommended to preserve marks and inscriptions
Specifications
Specifications
| Color | Transparent |
| Capacity (Metric) | 25 mL |
| Material | PMP |
| Height (Metric) | 108 mm |
| Autoclavable | Autoclavable |
| Sterility | Non-sterile |
| Class | B |
| Closure Type | Stopper |
| Closure Size | 10/19 |