Learn More
3M™ Plaster Roll
Product Code. 12133801 Shop All 3M Products
Change view
Click to view available options
Length: 9m
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
12133801 Length: 9m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12133801 Supplier 3M™ Supplier No. 7000032744
  • ONLINE EXCLUSIVE PRICE
    102.71€
    71.88€ / Pack of 12
    Save 30.83€
    30% Off
 Restricted Item
Estimated Shipment: 28-12-2026