Learn More
Plas Labs™ Rodent Injection Cone
Product Code. 11897132
Change view
Click to view available options
Color:
Clear
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
11897132 Clear Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11897132 Supplier Plas Labs™ Supplier No. 561RC
  • ONLINE EXCLUSIVE PRICE
    555.77€
    438.60€ / Pack of 1
    Save 117.17€
    21% Off
 Restricted Item
Estimated Shipment: 04-12-2026
Ideal for large quantity sampling which requires high speed and efficiency
  • Completely transparent, durable, and easy to clean
  • Utilizes suction cup feet to secure device
Specifications
Color Clear
For Use With (Application) Ideal for large quantity sampling requires high speed and efficiency
Type Rodent Injection Cone
Material Acrylic
Sterilization Method Chemically sterilized or washed at 180°F
Length (English) 5.75 in.
Length (Metric) 14.6 cm
Height (English) 5 in.
Height (Metric) 12.7 cm
Diameter (English) 4 in.
Diameter (Metric) 10.16 cm
Weight Range 15 to 30 g (Mice), 500 to 600 g (Rats)