Learn More
Plas Labs™ Flat-Bottom Rodent Restrainers
Product Code. 11807142
Change view
Click to view available options
Diameter (English):
1.5 in.
3.25 in.
Length (English):
3.75 in.
8 in.
Weight Range:
15 to 30 g
250 to 500 g
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (English) Length (English) Weight Range unitSize
11807142 1.5 in. 3.75 in. 15 to 30 g Each
11827142 3.25 in. 8 in. 250 to 500 g Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11807142 Supplier Plas Labs™ Supplier No. 541RR
Estimated Shipment: 18-08-2026
Add to basket
Add to basket
Apertures in the top and bottom provide easy access for injections or sampling
  • Pocket and holes in the front make it easy to anesthetize or feed the animal
  • Clear material provides a visual check of rodent's condition
  • Adjustable tail stock can be inserted into any one of four positions
Specifications
Color Clear
For Use With (Application) Injections or Blood Sampling
Type Flat Bottom Rodent Restrainer
Material Noryl™ Plastic
Sterilization Method Chemically sterilized or washed at 180°F
Length (English) 3.75 in.
Length (Metric) 9.52 cm
Diameter (English) 1.5 in.
Diameter (Metric) 3.81 cm
Weight Range 15 to 30 g