Learn More
Plas Labs™ Broome-Style Rodent Restrainers
Click to view available options
Diameter (English):
1 in.
2 in.
2.5 in.
Length (English):
3.75 in.
8 in.
8.5 in.
Weight Range:
125 to 250 g
15 to 30 g
250 to 500 g
Unit Size:
Each
Description
- Offer easy access to rodent
- Prevent rodent from turning
- Provide safety to the animal handler
- Manufactured from clear materials to provide complete visibility of animal
Specifications
Specifications
| Color | Clear |
| For Use With (Application) | Injections or Blood Sampling |
| Type | Rodent Restrainer |
| Material | Noryl™ Plastic |
| Sterilization Method | Chemically sterilized or washed at 180°F |
| Style | Broome |
| Length (English) | 8 in. |
| Length (Metric) | 20.32 cm |
| Diameter (English) | 2 in. |
| Diameter (Metric) | 5.08 cm |
| Show More |