Learn More
Olympus™ Plain Stage Insert Plate
Product Code. 12576965
Change view
Click to view available options
For Use With (Equipment):
IX-SP Microscopes
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
12576965 IX-SP Microscopes Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12576965 Supplier Olympus™ Supplier No. K10017352
Use to fulfill common microscope stage needs. Olympus™ Plain Stage Insert Plate is an exchangeable insert plate stage.
  • Exchangeable.
Specifications
Diameter (Metric) 110 mm
Dimensions (L x W) 232 x 240 mm
For Use With (Equipment) IX-SP Microscopes