Learn More
BRAND™ Plain Interior Polypropylene Funnels
Product Code. 15281459 Shop All Brand Products
Click to view available options
Diameter (Metric)Top:
50mm
75mm
100mm
120 mm
150mm
Height (Metric):
43 mm (Stem)
55 mm stem
77 mm (Stem)
90 mm (Stem)
95 mm (Stem)
Unit Size:
Pack of 12
Pack of 24
Product Code. 15281459
Brand: BRAND™ 147015
Edge
An excellent option with high temperature resistance for general laboratory use.
  • Available in a range of sizes
  • Sold in various quantities
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric)Top 50mm
Diameter (Metric) Stem 9mm
Material Polypropylene
Height (Metric) 43 mm (Stem)
Autoclavable Autoclavable
Length (Metric) Stem 43mm
Quantity 24/Pk.
Product Type Filling Funnel