Learn More
Abnova™ PLA2G4C Recombinant Protein
Brand:  Abnova™ H00008605-P01.10ug
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16162652
  • 359.00€ / 10µg
Estimated Shipment: 24-08-2022
Description
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifications
Specifications
AAH17956
Solution
8605
PLA2G4C (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
PLA2G4C
Human
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
48
8
Glutathione Sepharose 4 Fast Flow
10 ug
MGSSEVSIIPGLQKEEKAAVERRRLHVLKALKKLRIEADEAPVVAVLGSGGGLRAHIACLGVLSEMKEQGLLDAVTYLAGVSGSTWAISSLYTNDGDMEALEADLKHRFTRQEWDLAKSLQKTIQAARSENYSLTDFWAYMVISKQTRELPESHLSNMKKPVEEGTLPYPIFAAIDNDLQPSWQEARAPGKQTFRGRER
CPLA2-gamma/DKFZp586C0423
PLA2G4C