Learn More
Hecht Karl™ Piston Rings
Product Code. 10135331
Change view
Click to view available options
Size:
10 to 100 mL
2000 mL
250 to 1000 mL
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
10135331 2000 mL Each
10499401 10 to 100 mL Each
10227621 250 to 1000 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10135331 Supplier Hecht Karl™ Supplier No. 41914003
  • ONLINE EXCLUSIVE PRICE
    8.40€
    6.43€ / Pack of 1
    Save 1.97€
    23% Off
View all promotions
 Restricted Item
Estimated Shipment: 11-12-2026
Specifications
Material Cork
For Use With (Equipment) Round Bottom Flasks
No. per Pack 1 Each
Quantity 1 Each
Size 2000 mL
Product Type Piston Ring