Learn More
Pirimicarb-d6, TRC
Specifications
Specifications
| Chemical Name or Material | Pirimicarb D6 (dimethylcarbamate D6) |
| CAS | 1015854-66-6 |
| Molecular Formula | C11 2H6 H12 N4 O2 |
| Quantity | 2.5 mg |
| Synonym | Carbamic acid','N,N-di(methyl-d3)-','2-(dimethylamino)-5,6-dimethyl-4-pyrimidinyl ester (ACI), 2-(Dimethylamino)-5,6-dimethyl-4-pyrimidinyl N,N-di(methyl-d3)carbamate (ACI), Pirimicarb-d6, N,N-Di(methyl-d3)carbamic acid 2-(dimethylamino)-5,6-dimethyl-4-pyrimidinyl ester, Pirimicarb D6 (dimethylcarbamate D6) |
| InChI Formula | InChI=1S/C11H18N4O2/c1-7-8(2)12-10(14(3)4)13-9(7)17-11(16)15(5)6/h1-6H3/i5D3,6D3 |
| SMILES | [2H]C([2H])([2H])N(C(=O)Oc1nc(nc(C)c1C)N(C)C)C([2H])([2H])[2H] |
| IUPAC Name | [2-(dimethylamino)-5,6-dimethylpyrimidin-4-yl] N,N-bis(trideuteriomethyl)carbamate |
| Molecular Weight (g/mol) | 244.32 |
| Formula Weight | 244.1806 |