missing translation for 'onlineSavingsMsg'
Learn More

Pirbuterol Dihydrochloride, TRC

Product Code. 30133309
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133309 100 mg 100mg
30006792 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30133309 Supplier Toronto Research Chemicals Supplier No. TRCP502500100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38029-10-6
Molecular Formula C12 H20 N2 O3 . 2 Cl H
Molecular Weight (g/mol) 313.22
Synonym 2,6-Pyridinedimethanol, α6-[[(1,1-dimethylethyl)amino]methyl]-3-hydroxy-, hydrochloride (1:2), 2,6-Pyridinedimethanol, α6-[[(1,1-dimethylethyl)amino]methyl]-3-hydroxy-, dihydrochloride (9CI), Broncocor, CP 24314-1, Exirel, Exirel (bronchodilator), NSC 355078, Pirbuterol dihydrochloride, Pirbuterol hydrochloride, Pyrbuterol hydrochloride
IUPAC Name 6-[2-(tert-butylamino)-1-hydroxyethyl]-2-(hydroxymethyl)pyridin-3-ol;dihydrochloride
SMILES Cl.Cl.CC(C)(C)NCC(O)c1ccc(O)c(CO)n1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C12H20N2O3.2ClH/c1-12(2,3)13-6-11(17)8-4-5-10(16)9(7-15)14-8;;/h4-5,11,13,15-17H,6-7H2,1-3H3;2*1H
Formula Weight 312.1007
Chemical Name or Material Pirbuterol Dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.