Learn More
Hamilton™ 50μL Conductive CO-RE Pipet Tips in Nested Racks
Product Code. 10332165
Change view
Click to view available options
Volume (Metric):
50 μL
Unit Size:
Pack of 11520
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
10332165 50 μL Pack of 11520
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10332165 Supplier Hamilton™ Supplier No. 235947_CORE_I
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Essential for precise pipetting. Hamilton™ 50μL Conductive CO-RE Pipet Tips in Nested Racks provide enhanced pipetting safety and reliability with the CO-RE PerfectTouch system.
  • Usable with CO-RE 384 Probe Head
  • Fine, thin tip facilitates dosing of micro volumes
  • Capacitive LLD (cLLD) with conductive (black) version
  • Process safety offered by Monitored Air Displacement
Specifications
Volume (Metric) 50 μL
Filtered Non-filtered
Sterility Non-sterile
Disposable Single-use
For Use With (Equipment) CO-RE 384 Probe Head