Learn More
Hamilton™ 50μL Conductive CO-RE Pipet Tips in Nested Racks
Description
- Usable with CO-RE 384 Probe Head
- Fine, thin tip facilitates dosing of micro volumes
- Capacitive LLD (cLLD) with conductive (black) version
- Process safety offered by Monitored Air Displacement
Specifications
Specifications
| Volume (Metric) | 50 μL |
| Filtered | Non-filtered |
| Sterility | Non-sterile |
| Disposable | Single-use |
| For Use With (Equipment) | CO-RE 384 Probe Head |