Learn More
VistaLab™ Technologies Pipetter Nozzles
Product Code. 11936052
Click to view available options
Volume (Metric):
0.5 to 20 μL
100 to 200 μL
2 μL
2 to 20 μL
5 mL
Unit Size:
Pack of 10
Pack of 2
Pack of 4
Pack of 5
Product Code. 11936052
Brand: VistaLab™ Technologies 90605010
Contains an internal aerosol/liquid barrier filter to prevent liquid from being aspirated into pipetter
  • Available in range of sizes
Specifications
For Use With Pipettes
Material Plastic
Sterility Non-sterile
Volume (Metric) 5 mL
For Use With (Equipment) 100 to 5000 μL Macro Pipette
Quantity 2/Box
Length (English) 5 in.
Width (English) 3 in.
Height (English) 1 in.