Learn More
Eppendorf™ Pipette Holder 2 for one Xplorer™ Xplorer™ plus Pipette
GREENER_CHOICE
Product Code. 15779488 Shop All Eppendorf Products
Change view
Click to view available options
For Use With (Equipment):
Xplorer™ Xplorer™ plus Pipette
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
15779488 Xplorer™ Xplorer™ plus Pipette Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15779488 Supplier Eppendorf™ Supplier No. 3116000120
Only null left
Add to basket
Add to basket
Holder designed for wall mounting or for installation in Pipette Carousel 2
  • Carries one Eppendorf Xplorer™ or Xplorer™ plus
  • Same as Charger Shell 2 for Eppendorf Xplorer or Xplorer plus but without charging functionality
  • Designed for wall mounting or for installation in Pipette Carousel 2
  • Sticky tape for wall mounting included
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Xplorer™ Xplorer™ plus Pipette
Includes Sticky tape for wall mounting included