Learn More
Ohaus™ Pipette Adjustment Kit, EX Semi-Micro
Shop All Ohaus Products
Click to view available options
:
Compatible with EX model electronic balances
Description
- Compatible with EX model electronic balances
- EX125DM
- EX125M
- EX225DM
- EX225DM/AD
- EX225M/AD
- EX125D
- EX125
- EX225D
- EX225D/AD
- EX225/AD
Specifications
Specifications
| Description | Pipette Adjustment Kit, EX Semi-Micro |
| Type | Pipette Adjustment Kit, EX semi-micro |
| For Use With (Equipment) | Compatible with EX model electronic balances |