missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Piperidine-d11, CDN
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | Piperidine-D11 |
| CAS | 143317-90-2 |
| Molecular Formula | C5 2H11 N |
| Quantity | 1 g |
| Synonym | Piperidine-d11 (9 CI), Piperidine-D11 |
| InChI Formula | InChI=1 S/C5H11N/c1-2-4-6-5-3-1/h6H,1-5H2/i1D2,2D2,3D2,4D2,5D2/hD |
| SMILES | [2 H]N1C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C1([2 H])[2 H] |
| IUPAC Name | 1,2,2,3,3,4,4,5,5,6,6-undecadeuteriopiperidine |
| Molecular Weight (g/mol) | 96.215 |
| Formula Weight | 96.158 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction