missing translation for 'onlineSavingsMsg'
Learn More

Piperazine Dihydrochloride, TRC

Product Code. 30412795
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30412795 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30412795 Supplier Toronto Research Chemicals Supplier No. TRCP48010025G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Piperazine Dihydrochloride
CAS 142-64-3
Molecular Formula C4 H10 N2 . 2 Cl H
Quantity 25 g
Synonym Piperazine Dihydrochloride, Ascarex D, Diethylenediamine Dihydrochloride, Dihydro Pip Wormer, Dowzene DHC, Piperazine Hydrochloride, Piperazine Wormer Premix, Piperazinium Dichloride
InChI Formula InChI=1S/C4H10N2.2ClH/c1-2-6-4-3-5-1;;/h5-6H,1-4H2;2*1H
SMILES Cl.Cl.C1CNCCN1
IUPAC Name piperazine;dihydrochloride
Molecular Weight (g/mol) 159.06
Formula Weight 158.0378
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.