Learn More
Brady™ Pipe Markers - with Holder Water
Product Code. 12549747 Shop All Brady Products
Change view
Click to view available options
Display Text:
Eau de chauffe
Eau glacée
Eau pluviale
Unit Size:
Pack of 4
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Display Text Unit Size
12549747 Eau pluviale Pack of 4
12519747 Eau glacée Pack of 4
12539747 Eau de chauffe Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12549747 Supplier Brady™ Supplier No. N003425
  • ONLINE EXCLUSIVE PRICE
    15.65€
    14.14€ / Pack of 4
    Save 1.51€
    9% Off
Estimated Shipment: 24-11-2026
Specifications
Certifications/Compliance ROHS Environmental Compliance, EU Directive 2002/95/EC
Description Pipe Markers - Eau pluviale
Adhesive Type Permanent Acrylic
Color White on Green
Finish Gloss
Material Polyester
Product Type Pipe Marker
Display Text Eau pluviale
For Use With (Application) Pipe and valve marking
Labels per Sheet 4
Languages French
No. per Pack 4 Markers/Card
Quantity 4 Pk.
Temperature Range (English) -40°F to +230°F
Temperature Range (Metric) -40°C to +110°C
Thickness (English) 0.0039 in.
Thickness (Metric) 0.099 mm
Dimensions (L x W) 26 x 250 mm
Length (English) 1.02 in.
Length (Metric) 26 mm
Width (English) 9.84 in.
Width (Metric) 250 mm
Alternate Part Number EAU PLUVIALE 250X26CARD-T1-P16