Learn More
Thermo Scientific™ Piko PCR Plate, 96-well, white
Shop All Thermo Scientific Products
Click to view available options
Quantity:
200/Cs.
50 Plates
Unit Size:
Pack of 200
Pack of 50
Description
Piko PCR Plates are a family of PCR plates which have the footprint of a microscope slide. We have not only shrunk the size of the plate, but dramatically reduced the thickness of the well walls utilizing our patented ultra-thin wall (UTW) technology.
Four plates can be snapped into a single frame producing the equivalent of a standard 384-well microplate.
Features
• Ultra Thin Wall (UTW) for fast PCR/qPCR applications
• Low profile
• Designed for use with the Piko and PikoReal 96-well thermal cyclers
• Plates can be snapped into the plate frame to create a standard 384-well plate
• Compatible with standard multi-channel pipettes and liquid-handling platforms
• Well spacing and footprint conform to industry (ANSI) dimensions
Applications
• qPCR/Real-Time PCR
Related Products
Piko PCR Plate, 96-well, clear
Piko PCR Plate, 96-well, white
Plate Frame for 96-well Piko PCR Plates, white
Features
• Ultra Thin Wall (UTW) for fast PCR/qPCR applications
• Low profile
• Designed for use with the Piko and PikoReal 96-well thermal cyclers
• Plates can be snapped into the plate frame to create a standard 384-well plate
• Compatible with standard multi-channel pipettes and liquid-handling platforms
• Well spacing and footprint conform to industry (ANSI) dimensions
Applications
• qPCR/Real-Time PCR
Related Products
Piko PCR Plate, 96-well, clear
Piko PCR Plate, 96-well, white
Plate Frame for 96-well Piko PCR Plates, white
Specifications
Specifications
| Barcode | No Barcode |
| Certifications/Compliance | Certified RNase/DNase-free |
| Packaging | 50 Plate |
| Product Line | Piko |
| Color | White |
| Product Type | PCR Microplate |
| For Use With (Equipment) | Piko Thermal Cycler, PikoReal Real-Time PCR System |
| Material | Polypropylene |
| No. of Wells | 96 |
| Quantity | 50 Plates |
| Show More |