missing translation for 'onlineSavingsMsg'
Learn More

Pigment Red 22 (Technical Grade), TRC

Product Code. 30410573
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30410573 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30410573 Supplier Toronto Research Chemicals Supplier No. TRCP43778825MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Pigment Red 22
CAS 6448-95-9
Molecular Formula C24 H18 N4 O4
Quantity 25 mg
Synonym 2-Naphthalenecarboxamide','3-hydroxy-4-[(2-methyl-5-nitrophenyl)azo]-N-phenyl- (9CI), C.I. Pigment Red 22 (7CI,8CI), 3-Hydroxy-4-[2-(2-methyl-5-nitrophenyl)diazenyl]-N-phenyl-2-naphthalenecarboxamide, 3117 Bright Red, Accosperse Naphthol Scarlet, Alkali Re
InChI Formula InChI=1S/C24H18N4O4/c1-15-11-12-18(28(31)32)14-21(15)26-27-22-19-10-6-5-7-16(19)13-20(23(22)29)24(30)25-17-8-3-2-4-9-17/h2-14,29H,1H3,(H,25,30)
SMILES Cc1ccc(cc1N=Nc2c(O)c(cc3ccccc23)C(=O)Nc4ccccc4)[N+](=O)[O-]
IUPAC Name 3-hydroxy-4-[(E)-(2-methyl-5-nitrophenyl)diazenyl]-N-phenylnaphthalene-2-carboxamide
Molecular Weight (g/mol) 426.424
Formula Weight 426.133
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.