missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Ubiquitin Enrichment Kit

Product Code. 10158023 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
15 reactions kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10158023 15 reactions kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10158023 Supplier Thermo Scientific™ Supplier No. 89899

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Purify (enrich) poly-ubiquitinated proteins from cell and tissue lysates for post-translational modification studies using this affinity-resin, spin-column kit.

Thermo Scientific™ Pierce Ubiquitin Enrichment Kit facilitates the isolation of polyubiquitin protein conjugates from cultured cells and tissue samples.

The kit uses anti-ubiquitin affinity resin to bind and purify polyubiquitinated proteins (i.e., those with modifications containing more than four ubiquitin monomers) from cell lysates and other protein samples. The enriched fraction can subsequently be analyzed to determine the amount of general ubiquitin conjugates present or to identify a specific protein of interest by Western blotting. The assay protocol is fast and straight-forward, allowing for isolation of polyubiquitinated proteins and the fractionation of monoubiquitinated species in the resin flow-through. The Ubiquitin Enrichment Kit outperforms kits from other suppliers and provides a clean and specific preparation of proteins when compared to a control resin.

Highlights:

  • Fast – less than 45 minutes hands-on time

  • Complete – includes all reagents needed for ubiquitin-modified protein enrichment from cultured cells and tissue samples, as well as anti-ubiquitin antibody serum for Western blot analysis

  • Flexible – sample incubation from two hours to overnight allows assay to be completed in several hours or in less than 30 minutes after overnight incubation

  • Robust – compatible with all Thermo Scientific Pierce Cell Lysis Products and standard RIPA formulations

  • Multiple-sample format – easily processes 1 to 15 samples concurrently

Includes:

Polyubiquitin affinity resin (300μL), Tris-buffered saline pack, microcentrifuge spin columns and accessories, polyubiquitin positive control and anti-ubiquitin rabbit anti-serum

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Sufficient For: Enriching polyubiquitinated proteins from 15 lysate samples, each containing 0.15 mg total protein
• Polyubiquitin Positive Control (1000X), 50 μL (store at -20°C)
• Anti-ubiquitin Antibody (rabbit antiserum), 5 μL (store at -20°C)
• Polyubiquitin Affinity Resin Slurry, 300 μL (store at 4°C—do not freeze)
• TBS Pack (makes 500mL), 1 pack (store at 4°C)
• Spin Columns and Accessories, 18 columns (store at 4°C)
Product Line Pierce
Quantity 15 reactions kit
Target Polyubiquitinated Protein
Type Ubiquitin Enrichment Kit
Stationary Phase Proprietary Ligand
Sufficient For 15 Reactions
Format Kit, Microcentrifuge, Spin Column
Column Type Affinity, Agarose Resin
compliance-icons
Product Identifier
  • Pierce Ubiquitin Enrichment Kit
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: sodium chloride, trometamol

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.