Learn More
Thermo Scientific™ Pierce™ Phosphatase Inhibitor Mini Tablets
Product Code. p-7078345 Shop All Thermo Scientific Products
Click to view available options
Quantity:
20 Tablets
Unit Size:
Pack of 20
For Research Use Only. Not for use in diagnostic procedures.
Product Code. 15691759
Brand: Thermo Scientific™ A32957
For Research Use Only. Not for use in diagnostic procedures.
Thermo Scientific™ Pierce Phosphatase Inhibitor Mini Tablets provide complete cell lysate and biological sample protection from dephosphorylation with a broad spectrum phosphatase inhibitor formulation.
Pierce Phosphatase Inhibitor Mini Tablets protect phosphoproteins from serine/threonine phosphatases and tyrosine phosphatases. Each Phosphatase Inhibitor Tablet contains sodium fluoride, sodium orthovanadate, sodium pyrophosphate and beta-glycerophosphate for broad-spectrum inhibition of phosphatase activity. Tablets are stable at 4°C for up to 12 months, and each tablet is sufficient for 10mL of solution. The Pierce Phosphatase Inhibitor Tablets are suitable for use with any cell or tissue extract to preserve protein phosphorylation for downstream applications.
Highlights:
  • Economical&mdsashmore cost effective than other commercially available formulations
  • Improved formulation&mdsashtablets dissolve quickly into a clear solution
  • Complete&mdsashthe tablets inhibit acid, alkaline, and protein phosphatases
  • Compatible&mdsashuse with homebrew or commercially available cell lysis reagents and Thermo Scientific Protein Assays
TRUSTED_SUSTAINABILITY
Specifications
Product Type Protease Inhibitor Cocktail
Content And Storage 1 Vial, Store at 2°C to 8°C
Shipping Condition Wet Ice
Form Solid
Product Line Pierce
Sufficient For 10 mL Solution
Inhibitors Phosphatase Inhibitor
Quantity 20 Tablets
Composition Sodium Fluoride, Sodium Orthovanadate, Sodium Pyrophosphate, Beta-Glycerophosphate