Learn More
Description
Pierce Maleic Anhydride Activated Plates are useful for immobilizing peptides and other ligands that do not coat efficiently by passive adsorption. Reaction of the maleic anhydride groups with primary amines (–NH2) results in formation of amide bonds that are stable at neutral pH and above. Acidic conditions hydrolyze the bond; therefore, coating reactions are best performed at pH 8 to 9, and the reacted plates are best used at pH greater than 7 for ELISA and other methods.
These Thermo Scientific Pierce Maleic Anhydride Activated 96-Well Plates are clear, 96-well plates that allow attachment of amine-containing peptides and other molecules to microplate wells for use in binding assays.
Features of Maleic Anhydride Activated Plates:
• Spontaneously react with primary amines
• A direct, simple approach to covalently attach molecules to the well surface of an EIA plate
• Maleic anhydride maintains its integrity and availability for coupling for months under dry, room temperature conditions
Specifications
Specifications
| Binding Property | Standard Capacity, Covalent |
| Format | Standard |
| Content And Storage | Upon receipt store plates at room temperature. Product shipped at ambient temperature. |
| Detection Method | Colorimetric |
| Product Line | Pierce |
| Color | White |
| Product Type | Detection Plate, Immunoassay, ELISA |
| Material | Polystyrene |
| No. of Wells | 96 |
| Surface Treatment | Maleic Anhydride |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.