Learn More
Thermo Scientific™ Pierce™ Luciferase Cell Lysis Buffer (2X)
Product Code. 12142250 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12142250 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12142250 Supplier Thermo Scientific™ Supplier No. 16189
 Restricted Item
Estimated Shipment: 06-10-2026
Add to basket
Add to basket
Lyse adherent cells or pelleted cell suspensions for analysis of promoter expression in luciferase assays with this optimized protein extraction reagent.
Thermo Scientific™ Pierce Luciferase Cell Lysis Buffer effectively lyses adherent cells or pelleted suspension cells for analysis of promoter expression in luciferase assays.
Pierce Luciferase Cell Lysis Buffer is effective for preparing high-yield mammalian cell lysates from either adherent cells or pelleted suspension cells for analysis in luciferase reporter assays. The reagent is compatible with all Thermo Scientific Pierce Luciferase Assay Kits. The cell lysis buffer has been validated for use with COS-7, NIH3T3, Jurkat and other cell types. Lysates are produced rapidly and easily for luciferase assays and are also compatible with BCA and Coomassie (Bradford) protein assays.
Highlights:
  • Compatible with all Pierce Luciferase Reporter Assays
  • Enables rapid and efficient lysis
  • Tested compatibility with multiple Mammalian cell lines
  • Compatible with protein assays (e.g., Coomassie Plus and BCA)
TRUSTED_SUSTAINABILITY
Specifications
Type Cell Lysis Buffer
Quantity 250 mL
Content And Storage Store at room temperature.
Format Liquid