Learn More
Thermo Scientific™ Pierce™ Intact Protein Standard Mix
Product Code. 15776003 Shop All Thermo Scientific Products
Click to view available options
Quantity:
1 x 76 μg
5 x 76 μg
Unit Size:
76µg
Pack of 5
Product Code. 15776003
Brand: Thermo Scientific™ A33526
Edge
Specifications
For Use With (Equipment) Mass Spectrometer
Product Type Intact Protein Standard Mix
Product Line Pierce™
Content And Storage 1 vial, store at -5 to -30°C
Shipping Condition Wet Ice
Final Product Type Intact Proteins
Workflow Step Calibration
Quantity 1 x 76 μg
Form Powder