Learn More
Thermo Scientific™ Pierce™ Immunostain Enhancer
Product Code. 11805015 Shop All Thermo Scientific Products
Click to view available options
Quantity:
2 mL
Unit Size:
2mL
Product Code. 11805015
Brand: Thermo Scientific™ 46645
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Increase signal intensity 3- to 12-fold and antibody dilution 5- to 20-fold beyond normal using both chromogenic or fluorescent IHC substrates.
Thermo Scientific™ Pierce Immunostain Enhancer enables significant antibody dilution (5- to 20-fold beyond normal) to increase signal intensity 3- to 12-fold in immunostaining applications with chromogenic or fluorescent substrates.
This ready-to-use enhancer for immunohistochemical staining does not add steps to the workflow because it is used to dilute the primary and secondary antibodies. The Pierce Immunostain Enhancer is compatible with fluorescence and chromogenic detection and routinely increases both signal intensity and detection sensitivity. Signal enhancement is antibody-dependent and typically ranges from 3- to 12-fold. Because of the strong signal enhancement, the Pierce Immunostain Enhancer reduces the amount of antibody required to achieve optimal detection.
Highlights:
  • Save precious antibody – Pierce Immunostain Enhancer allows the customer to use only a fraction of antibody to achieve the same signal as with conventional immunodetection
  • Convenience – simply replace your current antibody dilution buffer with Pierce Immunostain Enhancer (unlike other signal enhancement methods which require additional steps)
  • Increased signal intensity and sensitivity – provides 3- to 12- fold increase in signal intensity and sensitivity for improved visualization of the antigen of interest in cells and tissues
  • Improved specificity – significantly improves signal-to-noise ratio for poor quality and low affinity antibodies
  • Compatible – can be used with chromogenic and fluorescent detection methods
TRUSTED_SUSTAINABILITY
Specifications
Description Pierce Immunostain Enhancer
Quantity 2 mL
Detection Method Chromogenic, Fluorescence
Sufficient For 10 large (approx. 3cm2) tissue section slides