Learn More
Thermo Scientific™ Pierce™ Detergent Compatible Bradford Assay Reagent
Product Code. 15308096 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
15 Assays
Unit Size:
25mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15308096 15 Assays 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15308096 Supplier Thermo Scientific™ Supplier No. 23246S
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Pierce™ Detergent Compatible Bradford Assay Reagent
The Thermo Scientific™ Pierce™ Detergent Compatible Bradford Assay Reagent is a quick and ready-to-use modification of the well-known Bradford coomassie dye-binding, colorimetric method for total protein quantitation. Proprietary additives to the Bradford Reagent make it compatible with up to 1% or higher of detergents and lysis reagents that are commonly used in life science research, including Triton™ X-100 and NP-40.
  • Convenient—detergent-free standard curve
  • Flexible—compatible with samples both with and without detergent
  • Minimal sample—requires only 10μL for microplate procedure
  • Colorimetric—measure with a standard spectrophotometer or plate reader (595 nm)
  • Easy to use—single reagent, no working reagent preparation required
  • Fast—10 minute incubation at room temperature
  • Broad range—detects protein concentration in the ranges of 2 to 1500μg/mL
    Similar to the Bradford method, coomassie dye binds protein in an acidic medium causing an immediate shift in absorption maximum from 465 nm to 595 nm with a concomitant color change from green to blue. In addition, the assay is complete in just 10 minutes.
    The protein assay can be performed in either test tube or microplate format. The standard working range is 100-1500μg/mL with up to 1% detergent (or higher in some cases). Protein concentrations are estimated by reference to absorbances obtained for a series of standard protein dilutions, typically bovine serum albumin (BSA), which are assayed alongside the unknown samples. Because the color response with coomassie dye is non-linear with increasing protein concentration, a standard curve must be completed with each assay. Standards can be used directly without preparing them in the same detergent found in the test samples.
  • TRUSTED_SUSTAINABILITY
    Specifications
    Description Detergent Compatible Bradford Assay Reagent, 15 Test Tube Assays, Protein Quantitation Assay Assay Type, Colorimetric Detection Method, Absorbance, Solution-based Detection Technique, Used For Protein Assays and Analysis
    Formulation Liquid
    For Use With (Application) Protein assays and analysis
    Sample Volume 15 Assay(s)
    Content And Storage 4°C
    Product Type Pierce™ Detergent Compatible Bradford Assay Reagent
    Quantity 15 Assays