Learn More
Thermo Scientific™ Pierce™ Anti-GST Coated Plates, Clear, 8-Well Strip
Shop All Thermo Scientific ProductsDescription
Thermo Scientific™ Pierce Anti-GST Coated Plates are polystyrene microplates coated with mouse monoclonal anti-GST antibody and pre-blocked for immediate use.
Additional Information and Online Ordering
Highlights:
Antibody-based – plates are coated with mouse monoclonal anti-GST antibody
Sensitive – detects GST fusion proteins down to 7.5ng/well
Flexibility – binds native or denatured forms of GST; an alternative to glutathione-coated plates
Ready to use – pre-blocked with SuperBlock Blocking Buffer
Compatibility – Thermo Scientific Cell Lysis Reagents and many other popular lysis buffers do not interfere with GST-fusion protein binding
Superior performance – improved signal-to-noise ratio relative to competitor plates
Specifications
Specifications
| Binding Property | Standard Capacity, Affinity |
| Content And Storage | Store plates at 4°C. Store opened, unused plates in a resealable bag with desiccant at 4°C. Shipped at ambient temperature. |
| Detection Method | Colorimetric |
| Packaging | 5 Plate |
| Product Line | Pierce |
| Color | Clear |
| Product Type | Detection Plate, Immunoassay, ELISA |
| Material | Polystyrene |
| No. of Wells | 96 |
| Surface Treatment | Antibody |
| Show More |