Learn More
4-Fluorotoluene-alpha,alpha,alpha-d3, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
Specifications
Specifications
| Chemical Name or Material | 4-Fluorotoluene-alpha,alpha,alpha-D3 |
| CAS | 4202-91-9 |
| Molecular Formula | C7 D3 H4 F |
| Quantity | 0.1 g |
| Synonym | Benzene, 1-fluoro-4-(methyl-d3)- (9 CI, ACI), 1-Fluoro-4-(methyl-d3)benzene (ACI), Toluene-α,α,α-d3, p-fluoro- (7 CI, 8 CI), 1-Fluoro-4-(2H3)methylbenzene, 4-Fluorotoluene-α,α,α-d3, 4-Fluorotoluene-α,α,α-D3 |
| InChI Formula | InChI=1 S/C7H7F/c1-6-2-4-7(8)5-3-6/h2-5 H,1H3/i1D3 |
| SMILES | [2 H]C([2 H])([2 H])c1ccc(F)cc1 |
| IUPAC Name | 1-fluoro-4-(trideuteriomethyl)benzene |
| Molecular Weight (g/mol) | 113.1474 |
| Formula Weight | 113.072 g/mol |
| Show More |