missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tri-n-butyl(vinyl-d3)tin, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | Tributylvinyltin-D3 (vinyl-D3) |
| Molecular Formula | C14 D3 H27 Sn |
| Quantity | 0.5 g |
| Synonym | Tributylethenyl-d3-stannane, (Ethenyl-1,2,2-d3)tributyltin, (Tributylstannyl)ethene-1,2,2-d3, Tributyl(vinyl-d3)stannane, Tributylvinyltin-d3, Tributylvinyltin-D3 (vinyl-D3), (Vinyl-d3)tributyltin |
| InChI Formula | InChI=1 S/3C4H9.C2H3.Sn/c3*1-3-4-2;1-2;/h3*1,3-4H2,2H3;1 H,2H2;/i;;;1 D,2D2; |
| SMILES | [2 H]C(=C([2 H])[Sn](CCCC)(CCCC)CCCC)[2 H] |
| IUPAC Name | tributyl(1,2,2-trideuterioethenyl)stannane |
| Molecular Weight (g/mol) | 320.116 |
| Formula Weight | 321.156 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction