Learn More
Phthalimidoacetaldehyde, TRC
Specifications
Specifications
| Chemical Name or Material | Phthalimidoacetaldehyde |
| CAS | 2913-97-5 |
| Molecular Formula | C10 H7 N O3 |
| Quantity | 1 g |
| InChI Formula | InChI=1S/C10H7NO3/c12-6-5-11-9(13)7-3-1-2-4-8(7)10(11)14/h1-4,6H,5H2 |
| SMILES | O=CCN1C(=O)c2ccccc2C1=O |
| IUPAC Name | 2-(1,3-dioxoisoindol-2-yl)acetaldehyde |
| Molecular Weight (g/mol) | 189.17 |
| Formula Weight | 189.0426 |