missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phthalic acid, n-butyl-n-decyl ester 100 μg/mL in Hexane, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | Phthalic Acid, n-Butyl-n-Decyl Ester |
| CAS | 89-19-0 |
| Molecular Formula | C22 H34 O4 |
| Quantity | 1 mL |
| Synonym | Phthalic Acid, n-Butyl-n-Decyl Ester |
| InChI Formula | InChI=1S/C22H34O4/c1-3-5-7-8-9-10-11-14-18-26-22(24)20-16-13-12-15-19(20)21(23)25-17-6-4-2/h12-13,15-16H,3-11,14,17-18H2,1-2H3 |
| SMILES | CCCCCCCCCCOC(=O)c1ccccc1C(=O)OCCCC |
| IUPAC Name | 1-O-butyl 2-O-decyl benzene-1,2-dicarboxylate |
| Molecular Weight (g/mol) | 362.503 |
| Formula Weight | 362.2457 g/mol |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?