missing translation for 'onlineSavingsMsg'
Learn More

Phthalic acid monosodium 100 μg/mL in Water, Dr. Ehrenstorfer

Product Code. 30316422
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30316422 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30316422 Supplier Dr. Ehrenstorfer Supplier No. DREA16180000WA100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium Hydrogen Phthalate
CAS 827-27-0
Molecular Formula C8 H5 O4 . Na
Quantity 1 mL
Synonym Sodium Hydrogen Phthalate, 1,2-Benzenedicarboxylic acid, monosodium salt (9CI), Phthalic acid, monosodium salt (8CI), Monosodium phthalate, Sodium acid phthalate, Sodium hemiphthalate, Sodium hydrogen phthalate
InChI Formula InChI=1S/C8H6O4.Na/c9-7(10)5-3-1-2-4-6(5)8(11)12;/h1-4H,(H,9,10)(H,11,12);/q;+1/p-1
SMILES [Na+].OC(=O)c1ccccc1C(=O)[O-]
IUPAC Name sodium;2-carboxybenzoate
Molecular Weight (g/mol) 188.113
Formula Weight 188.0086 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.