Learn More
Phthalic Acid Mono-tetrahydrogeranyl Ester, TRC
Product Code. 30133133
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133133 1 g 1g
30005665 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30133133 Supplier Toronto Research Chemicals Supplier No. TRCP3845401G
 Restricted Item
Estimated Shipment: 12-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 297182-84-4
Molecular Formula C18 H26 O4
Molecular Weight (g/mol) 306.4
Synonym Phthalic acid 3,7-Dimethyloctyl ester, Monoisodecyl phthalate
IUPAC Name 2-(3,7-dimethyloctoxycarbonyl)benzoic acid
SMILES CC(C)CCCC(C)CCOC(=O)c1ccccc1C(=O)O
Specifications
Quantity 1 g
InChI Formula InChI=1S/C18H26O4/c1-13(2)7-6-8-14(3)11-12-22-18(21)16-10-5-4-9-15(16)17(19)20/h4-5,9-10,13-14H,6-8,11-12H2,1-3H3,(H,19,20)
Formula Weight 308.1831
Chemical Name or Material 3,7-dimethyloctyl phthalate